missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | RPB9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630141
|
Novus Biologicals
NBP2-93676-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633470
|
Novus Biologicals
NBP2-93676-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPB9 Polyclonal antibody specifically detects RPB9 in Human, Mouse samples. It is validated for Western BlotSpecifications
| RPB9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 5438 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA-directed RNA polymerase II 14.5 kDa polypeptide, DNA-directed RNA polymerase II subunit I, DNA-directed RNA polymerase II subunit RPB9, hRPB14.5, polymerase (RNA) II (DNA directed) polypeptide I (14.5kD), polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa, RNA polymerase II 14.5 kDa subunit, RNA polymerase II subunit B9, RPB14.5, RPB9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human POLR2I (NP_006224.1). MEPDGTYEPGFVGIRFCQECNNMLYPKEDKENRILLYACRNCDYQQEADNSCIYVNKITHEVDELTQIIADVSQDPTLPRTEDHPCQKCGHKEAVFFQSHSARAEDAMRLYYVCTAPHCGHRWTE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title