missing translation for 'onlineSavingsMsg'
Learn More

RPB9 Antibody (2B3), Novus Biologicals™

Product Code. 18375349 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375349 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375349 Supplier Novus Biologicals Supplier No. H00005438M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPB9 Monoclonal antibody specifically detects RPB9 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPB9
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2B3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17112
Gene Alias DNA-directed RNA polymerase II 14.5 kDa polypeptide, DNA-directed RNA polymerase II subunit I, DNA-directed RNA polymerase II subunit RPB9, hRPB14.5, polymerase (RNA) II (DNA directed) polypeptide I (14.5kD), polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa, RNA polymerase II 14.5 kDa subunit, RNA polymerase II subunit B9, RPB14.5, RPB9
Host Species Mouse
Immunogen POLR2I (AAH17112, 26 a.a. ~ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PKEDKENRILLYACRNCDYQQEADNSCIYVNKITHEVDELTQIIADVSQDPTLPRTEDHPCQKCGHKEAVFFQSHSARAEDAMRLYYVCTAPHCGHRWTE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5438
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.