missing translation for 'onlineSavingsMsg'
Learn More

RPB2 Antibody (HL1042), Novus Biologicals™

Product Code. 18385815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385815 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385815 Supplier Novus Biologicals Supplier No. NBP313697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RPB2 Monoclonal antibody specifically detects RPB2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPB2
Applications Western Blot
Classification Monoclonal
Clone HL1042
Conjugate Unconjugated
Formulation PBS
Gene Alias DNA-directed RNA polymerase II subunit RPB2, EC 2.7.7, EC 2.7.7.6, hRPB140, hsRPB2, POL2RB, polymerase (RNA) II (DNA directed) polypeptide B (140kD), polymerase (RNA) II (DNA directed) polypeptide B, 140kDa, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RNA polymerase II subunit2, RNA polymerase II second largest subunit
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human RPB2. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5431
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.