missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93593-0.1ml
This item is not returnable.
View return policy
Description
RPB2 Polyclonal antibody specifically detects RPB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RPB2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| DNA-directed RNA polymerase II subunit RPB2, EC 2.7.7, EC 2.7.7.6, hRPB140, hsRPB2, POL2RB, polymerase (RNA) II (DNA directed) polypeptide B (140kD), polymerase (RNA) II (DNA directed) polypeptide B, 140kDa, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RNA polymerase II subunit2, RNA polymerase II second largest subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human POLR2B (NP_000929.1). MYDADEDMQYDEDDDEITPDLWQEACWIVISSYFDEKGLVRQQLDSFDEFIQMSVQRIVEDAPPIDLQAEAQHASGEVEEPPRYLLKFEQIYLSKPTHWERDGAPSPMMPNEARLRNLTYSAPLYVDITKTVIKEGEEQLQTQHQKTFIGKIPIMLRSTYCLLNGLTDRDLCELNECPLDPGGYFIINGSEKVLIAQEKMATNTVYVFAKKDSKYAYTGECRSCLENSSRPTSTIWVSMLARGGQGAKKSAIGQRIVATL | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 5431 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction