missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17401-1-AP-150UL
This item is not returnable.
View return policy
Description
RPAP2 belongs to the RNA Pol II-associated protein (RPAP) family, a set of proteins involved in human Pol II machinery. Recently it has been reported that RPAP2 is required for efficient transcription of snRNA genes by Pol II. siRNA-mediated knock down of RPAP2 caused defects in snRNA gene expression. (22137580).Specifications
| RPAP2 | |
| Polyclonal | |
| Unconjugated | |
| RPAP2 | |
| C1orf82, RPAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231571, 79871 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IXW5, Q8VC34 | |
| RPAP2 | |
| RPAP2 Fusion Protein Ag11374 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction