Learn More
RPAP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13248
This item is not returnable.
View return policy
Description
RPAP2 Polyclonal specifically detects RPAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RPAP2 | |
| Polyclonal | |
| RUO |
| Immunohistochemistry | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: RCSRKAAGTKQTSTLKQEDASKRKAELEAAVRKKIEFERKALHIVEQLLEENITEEFLMECGRFITPAHYSDVVDERSIVKLCGYPLCQKKLGI | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |