missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104116
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPAP1.
RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.
Specifications
| RPAP1 | |
| Polyclonal | |
| Unconjugated | |
| RPAP1 | |
| 1190005L06Rik; A730023M06Rik; AU041788; AW107702; Kiaa1403; mKIAA1403; RNA polymerase II associated protein 1; RNA polymerase II-associated protein 1; RPAP1; RPAP-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26015 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BWH6 | |
| RPAP1 | |
| A synthesized peptide derived from human RPAP1(Accession Q9BWH6), corresponding to amino acid residues E353-V403. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction