missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPAIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554928
This item is not returnable.
View return policy
Description
Immunogen sequence: YNLRITSGVV VCQCGLSIPS HSSELTEQKL RACLEGSINE HSAHCPHTPE FSVTGGTEEK SSLLMSCLAC DTWAVI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 84%.
Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
Specifications
| RPAIN | |
| Polyclonal | |
| Unconjugated | |
| RPAIN | |
| 2400006N03Rik; hRIP; nuclear transporter; RAP interaction protein; RAP interaction protein alpha; RGD1308492; RIP; RPA interacting protein; RPA interacting protein delta 2; RPA interacting protein epsilon; RPAIN; RPA-interacting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UA6 | |
| RPAIN | |
| Recombinant protein corresponding to Human RPAIN. Recombinant protein control fragment (Product #RP-93238). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction