missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAIN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | RPAIN |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18497590
|
Novus Biologicals
NBP1-81707-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18426110
|
Novus Biologicals
NBP1-81707 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPAIN Polyclonal antibody specifically detects RPAIN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RPAIN | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 84268 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ25625, FLJ30490, FLJ42429, hRIP, MGC4189, nuclear transporter, RAP interaction protein, RIPHRIP, RPA interacting protein, RPA-interacting protein | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YNLRITSGVVVCQCGLSIPSHSSELTEQKLRACLEGSINEHSAHCPHTPEFSVTGGTEEKSSLLMSCLACDTWAVI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title