missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529616
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (89%), Sheep (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
Specifications
| RPA70 | |
| Polyclonal | |
| Unconjugated | |
| RPA1 | |
| 5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6117 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P27694 | |
| RPA1 | |
| Recombinant fragment corresponding to a region within amino acids 239 and 566 of Human RPA70 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction