missing translation for 'onlineSavingsMsg'
Learn More

RPA2, p Ser4, p Ser8 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18414743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414743 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414743 Supplier Novus Biologicals Supplier No. NBP123017IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RPA2 Polyclonal antibody specifically detects RPA2 in Human, Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA2
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34
Host Species Rabbit
Immunogen Immunogen was a dually phosphorylated synthetic peptide, which represented a portion of human replication protein A2, 32 kDa surrounding phosphorylated serines that corresponded to positions 4 and 8 using the numbering given in entry NP_002937.1 (GeneID
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.