missing translation for 'onlineSavingsMsg'
Learn More

RPA2 Antibody (RPA2/4774), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18319075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319075 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319075 Supplier Novus Biologicals Supplier No. NBP314167AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RPA2/4774
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34
Host Species Mouse
Immunogen Recombinant human full-length RPA2protein (Uniprot: P15927)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.