missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RP9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570635
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 79%.
Is thought to be a target protein for the PIM1 kinase. May play some roles in B-cell proliferation in association with PIM1.
Specifications
| RP9 | |
| Polyclonal | |
| Unconjugated | |
| RP9 | |
| LOW QUALITY PROTEIN: retinitis pigmentosa 9 protein; PAP1; PAP-1; Pim-1 associated protein; Pim-1 kinase associated protein; pim-1-associated protein; retinitis pigmentosa 9 (autosomal dominant); retinitis pigmentosa 9 (human); retinitis pigmentosa 9 homolog; retinitis pigmentosa 9 protein; Retinitis pigmentosa 9 protein homolog; RGD1559759; RP9; RP9 pre-mRNA splicing factor; Rp9h | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6100 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TA86 | |
| RP9 | |
| synthetic peptide directed towards the middle region of human RP9 (aa 172-221). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction