missing translation for 'onlineSavingsMsg'
Learn More

RP5-1077B9.4 Antibody, Novus Biologicals™

Product Code. 18422869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422869 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422869 Supplier Novus Biologicals Supplier No. H00060672B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RP5-1077B9.4 Polyclonal antibody specifically detects RP5-1077B9.4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RP5-1077B9.4
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. AAH08068.1
Gene Alias FLJ12438, FLJ38609, IGFBP2-binding protein, IGFBP-2-Binding Protein, IIp45, invasion inhibitory protein 45, Invasion-inhibitory protein 45, migration and invasion inhibitory protein, migration and invasion-inhibitory protein
Host Species Mouse
Immunogen MIIP (AAH08068.1, 1 a.a. - 388 a.a.) full-length human protein. MVEAEELAQLRLLNLELLRQLWVGQDAVRRSVARAASESSLESSSSYNSETPSTPETSSTSLSTSCPRGRSSVWGPPDACRGDLRDVARSGVASLPPANCQHQESLGRPRPHSAPSLGTSSLRDPEPSGRLGDPGPQEAQTSRSILAQQSKLSKPRVTFSEESAVPERSWRLRPYLGYDWIAGSLDTSSSITSQPEAFFSKLQEFRETNKEECICSHPEPQLPGLRESSGSGVEEDHECVYCYRVNRRLFPVPVDPGTPCRLCRTPRDQQGPGTLAQPAHVRVSIPLSILEPPHRYHIHRRKSFDASDTLALPRHCLLGWDIFPPKSEKSSAPRNLDLWSSVSAEAQHQKLSGTSSPFHPASPMQMLPPTPTWSVPQVPRPHVPRQKP
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 60672
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.