missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP105/CD180 Antibody (G28-8), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62228APCCY7
This item is not returnable.
View return policy
Description
RP105/CD180 Monoclonal specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry.
Specifications
| RP105/CD180 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CD180 | |
| Human tonsillar B cells | |
| 0.1 mL | |
| Primary | |
| The antibody VI-01 reacts with vimentin, a 57 kDa intermediate filament expressed in variety of mesenchymal and mesodermal cell types. Cross-reactivity was found with smooth muscle desmin. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| G28-8 | |
| Flow Cytometry | |
| CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Immunology, Innate Immunity | |
| 4064.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction