missing translation for 'onlineSavingsMsg'
Learn More

RP105/CD180 Antibody (G28-8), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18413106 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413106 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413106 Supplier Novus Biologicals Supplier No. NBP262228AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RP105/CD180 Monoclonal antibody specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen RP105/CD180
Applications Flow Cytometry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone G28-8
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Host Species Mouse
Immunogen Human tonsillar B cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 4064
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.