missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559928
This item is not returnable.
View return policy
Description
Immunogen sequence: VTCSPCEMCT VNKAYSPKET CNPSDTFFPS DGYGVDQTSM NKACFLGEVC SLTDTVFSDK ACAQKENHTY EGACPIDETY VPVNVCNTID FLNSKENTYT DNLDSTEELE RGDDIQKDLN ILTDPEYKNG FNTLVSHQNV SNLSSCG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 49%.
Microtubule-associated protein regulating the stability and length of the microtubule-based axoneme of photoreceptors. Required for the differentiation of photoreceptor cells, it plays a role in the organization of the outer segment of rod and cone photoreceptors ensuring the correct orientation and higher-order stacking of outer segment disks along the photoreceptor axoneme.
Specifications
| RP1 | |
| Polyclonal | |
| Unconjugated | |
| RP1 | |
| Dcdc3; DCDC4A; mG145; Orp1; oxygen-regulated protein 1; retinitis pigmentosa 1 (autosomal dominant); retinitis pigmentosa 1 (human); retinitis pigmentosa 1 homolog; retinitis pigmentosa 1 protein; Retinitis pigmentosa RP1 protein; retinitis pigmentosa RP1 protein homolog; RP1; Rp1h | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56715 | |
| RP1 | |
| Recombinant protein corresponding to Human RP1. Recombinant protein control fragment (Product #RP-89748). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction