missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22149C
This item is not returnable.
View return policy
Description
RP1 Polyclonal specifically detects RP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RP1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| DCDC4A, FLJ50293, FLJ55454, oxygen-regulated protein 1, retinitis pigmentosa 1 (autosomal dominant), retinitis pigmentosa 1 protein, retinitis pigmentosa RP1 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RP1 | |
| A synthetic peptide made to a C-terminal portion of the human RP1 protein (between residues 2000-2156) [UniProt P56715] | |
| 0.1 mL | |
| Hypoxia, Neuroscience, Signal Transduction | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction