missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £395.00
Specifications
| Antigen | RP1 |
|---|---|
| Concentration | 2.0 mg/mL |
| Dilution | Western Blot 2.0 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18176045
|
Novus Biologicals
NBP2-22149SS |
0.025 mL |
£188.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18117871
|
Novus Biologicals
NBP2-22149 |
0.1 mL |
£395.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RP1 Polyclonal specifically detects RP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RP1 | |
| Western Blot 2.0 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:400 | |
| Polyclonal | |
| Rabbit | |
| Hypoxia, Neuroscience, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| DCDC4A, FLJ50293, FLJ55454, oxygen-regulated protein 1, retinitis pigmentosa 1 (autosomal dominant), retinitis pigmentosa 1 protein, retinitis pigmentosa RP1 protein | |
| RP1 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Predicted to react with monkey based upon 100% sequence homology. |
| 2.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P56715, P56716 | |
| 6101 | |
| A synthetic peptide made to a C-terminal portion of the human RP1 protein (between residues 2000-2156) [UniProt P56715] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 240 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title