missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP-1 Antigen Mouse anti-Rat , PE-CF594, Clone: RP-1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566866
This item is not returnable.
View return policy
Description
RP-1 Antigen Monoclonal antibody specifically detects RP-1 Antigen in Rat samples. It is validated for Flow Cytometry
Specifications
| RP-1 Antigen | |
| Monoclonal | |
| PE-CF594 | |
| RP1 Antigen; Rat Granulocytes; Rat Neutrophils | |
| WKA/Hok rat peritoneal neutrophils | |
| RUO | |
| Rat | |
| IgG2a κ |
| Flow Cytometry | |
| RP-1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction