missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP-1 Antigen Mouse anti-Rat , BB700, Clone: RP-1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566871
This item is not returnable.
View return policy
Description
RP-1 Antigen Monoclonal antibody specifically detects RP-1 Antigen in Rat samples. It is validated for Flow Cytometry
Specifications
| RP-1 Antigen | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| RP1 Antigen; Rat Granulocytes; Rat Neutrophils | |
| WKA/Hok rat peritoneal neutrophils | |
| RUO | |
| Rat | |
| IgG2a κ |
| Flow Cytometry | |
| RP-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction