missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP 001 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent agonist of S1P1 receptors (EC50 = 9 pM). Induces internalization and polyubiquitination of S1P1 in vitro. Selective for S1P1 over S1P2 - S1P4.
Specifications
Formula Weight | 468.93 |
Percent Purity | >98% |
Quantity | 10 mg |
Chemical Name or Material | RP 001 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'