missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RP 001 hydrochloride

Product Code. 18054632
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054632 50 mg 50mg
18044972 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054632 Supplier Tocris Bioscience™ Supplier No. 4289/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent S1P1 agonist

Potent agonist of S1P1 receptors (EC50 = 9 pM). Induces internalization and polyubiquitination of S1P1 in vitro. Selective for S1P1 over S1P2 - S1P4.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306761-53-4
Molecular Formula C24H25ClN4O4

Specifications

Quantity 50 mg
Formula Weight 468.93
Purity >98%
Molecular Weight (g/mol) 468.938
Chemical Name or Material RP 001 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.