missing translation for 'onlineSavingsMsg'
Learn More

Roxindole Hydrochloride, TRC

Product Code. 30418892
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418892 10 mg
30427711 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418892 Supplier Toronto Research Chemicals Supplier No. TRCR70085810MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108050-82-4
Molecular Formula C23H26N2O . (HCl)
Molecular Weight (g/mol) 382.9
Synonym 3-[4-(3,6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol Hydrochloride, 3-[4-(3,6-dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-Indol-5-ol Monohydrochloride, Roxindole Hydrochloride
IUPAC Name 3-[4-(4-phenyl-3,6-dihydro-2H-pyridin-1-yl)butyl]-1H-indol-5-ol;hydrochloride
SMILES Cl.Oc1ccc2[nH]cc(CCCCN3CCC(=CC3)c4ccccc4)c2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H26N2O.ClH/c26-21-9-10-23-22(16-21)20(17-24-23)8-4-5-13-25-14-11-19(12-15-25)18-6-2-1-3-7-18;/h1-3,6-7,9-11,16-17,24,26H,4-5,8,12-15H2;1H
Formula Weight 382.1812
Chemical Name or Material Roxindole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.