missing translation for 'onlineSavingsMsg'
Learn More

Roxatidine Acetate Hydrochloride, MedChemExpress

Product Code. 16452129
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16452129

Brand: Medchem Express HYB0305A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Roxatidine Acetate Hydrochloride is a potent, selective, competitive and orally active histamine H2-receptor antagonist. Roxatidine Acetate Hydrochloride has antisecretory potency against gastric acid secretion. Roxatidine Acetate Hydrochloride can also suppress inflammatory responses and can be used for gastric and duodenal ulcers research. Roxatidine Acetate Hydrochloride has antitumor activity.

Roxatidine Acetate Hydrochloride is a potent, selective, competitive and orally active histamine H2-receptor antagonist. Roxatidine Acetate Hydrochloride has antisecretory potency against gastric acid secretion. Roxatidine Acetate Hydrochloride can also suppress inflammatory responses and can be used for gastric and duodenal ulcers research. Roxatidine Acetate Hydrochloride has antitumor activity.

Specifications

Chemical Name or Material Roxatidine Acetate Hydrochloride
CAS 93793-83-0
Color White
Purity Grade Notes Research
Molecular Formula C19H29ClN2O4
Quantity 1 g
Synonym HOE 760
Solubility Information DMSO : 50 mg/mL (129.90 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (129.90 mM)
SMILES O=C(NCCCOC1=CC=CC(CN2CCCCC2)=C1)COC(C)=O.Cl
Molecular Weight (g/mol) 384.9
Formula Weight 384.9
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.