missing translation for 'onlineSavingsMsg'
Learn More

Rovazolac, MedChemExpress

Product Code. 16496987
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496987 5 mg 5mg
16495196 100 mg 100mg
16484268 10 mg 10mg
16400607 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496987 Supplier Medchem Express Supplier No. HY109073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rovazolac is a liver x receptor (LXR) modulator extracted from patent WO2013130892A1.

Rovazolac is a liver x receptor (LXR) modulator extracted from patent WO2013130892A1.

Especificaciones

Chemical Name or Material Rovazolac
CAS 1454288-88-0
Color White
Purity Grade Notes Research
Molecular Formula C21H19F3N2O4S
Quantity 5 mg
Solubility Information Ethanol : 25 mg/mL (55.25 mM; Need ultrasonic)
SMILES O=C(OCC)CN1C(C2=CC=C(C3=CC(S(C)(=O)=O)=CC=C3)C=C2)=CC(C(F)(F)F)=N1
Molecular Weight (g/mol) 452.45
Formula Weight 452.45
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.