missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Routine DNA Ladder

Product Code. 10450464
Click to view available options
No. of Bands:
11
16
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10450464

Brand: Fisher BioReagents™ BP2582200

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ready-to-use DNA size standard is room-temperature stable for up to two years

  • Includes a landmark doublet at 1400 and 1550 bp

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Product Type DNA Molecular Weight Marker
Form Liquid
For Use With (Application) General purpose, wide DNA size range
No. of Bands 16
Quantity 1000 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.