Learn More
Manutan Round sling, 2000kg capacity, green, 3m circumference
Product Code. 12838899
Encompass_Preferred
Product Code. 12838899 Supplier Manutan Supplier No. A340102
Flexible, round, cross-section fabric giving excellent grip on awkward shaped loads. 100% polyester, woven construction. Unaffected by sunlight, grease or rust; resistant to dry heat and some acids and alkalis. Safety factor 7:1, manufactured to BS EN 1492-2:2000. All slings are colour coded and individually numbered and labelled to indicate the Safe Working Load (SWL). Test certificate supplied with each order, CE94 approved.
  • Vertical lift SWL: 2000kg
  • Green