Learn More
IKA™ Round Bottom Flask Inlay
Round bottom flask inlay constructed of anodized aluminum
Brand:  IKA™ 0025000644
Additional Details : Weight : 0.92000kg
 View more versions of this product
Product Code. 15284037
  • £140.00 / Each
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Round bottom
  • Anodized aluminum construction
  • Temperature resistant
Specifications
Specifications
500 mL
108 mm
180°C
4.3 in.
Round Bottom
Flask Inlay