missing translation for 'onlineSavingsMsg'
Learn More

Rotundine, MedChemExpress

Product Code. 16474747
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16474747 Supplier Medchem Express Supplier No. HYN0096
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rotundine is an antagonist of dopamine D1, D2 and D3 receptors with IC50s of 166 nM, 1.4 μM and 3.3 μM, respectively. Rotundine is also an antagonist of 5-HT1A with an IC50 of 370 nM.

Rotundine is an antagonist of dopamine D1, D2 and D3 receptors with IC50s of 166 nM, 1.4 μM and 3.3 μM, respectively. Rotundine is also an antagonist of 5-HT1A with an IC50 of 370 nM.

Specifications

Chemical Name or Material Rotundine
CAS 483-14-7
Color White
Purity Grade Notes Research
Molecular Formula C21H25NO4
Quantity 50 mg
Synonym (-)-Tetrahydropalmatine L-Tetrahydropalmatine
Solubility Information DMSO : ≥ 100 mg/mL (281.35 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC=C2C(CN3CCC4=CC(OC)=C(OC)C=C4[C@]3([H])C2)=C1OC
Molecular Weight (g/mol) 355.43
Formula Weight 355.43
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.