Learn More
SVL™ Rotulex™ Spherical Joints
Click to view available options
Standard Taper Joint No.:
13
19
29
35
41
64
Includes:
Female Joint
Female Joint, PTFE Covered O-ring, Spring-loaded Metal clip
Male Joint
Male Joint, PTFE Covered O-ring, Spring-loaded Metal clip
Unit Size:
Pack of 20
Pack of 30
Pack of 5
Description
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
- Joint can be retained together with spring loaded metal clip
- Component parts RM, RF, RO and RC should be ordered separately
Specifications
Specifications
| Includes | Female Joint |
| Material | Glass, PTFE |
| Product Type | Spherical joint |
| Standard Taper Joint No. | 19 |