missing translation for 'onlineSavingsMsg'
Learn More

Rottlerin, TRC

Product Code. 30414186
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414186 10 mg 10mg
30409956 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414186 Supplier Toronto Research Chemicals Supplier No. TRCR70074010MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-08-6
Molecular Formula C30 H28 O8
Molecular Weight (g/mol) 516.54
Synonym 3'-((8-Cinnamoyl-5,7-dihydroxy-2,2-dimethyl-2H-1-benzopyran-6-yl)methyl)-2',4',6'-trihydroxy-5'-methyl-acetophenone, Mallotoxin
IUPAC Name (E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methyl-phenyl)methyl]-5,7-dihydroxy-2,2-dimethyl-chromen-8-yl]-3-phenyl-prop-2-en-1-one
SMILES CC(=O)c1c(O)c(C)c(O)c(Cc2c(O)c3C=CC(C)(C)Oc3c(C(=O)\C=C\c4ccccc4)c2O)c1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C30H28O8/c1-15-24(33)19(27(36)22(16(2)31)25(15)34)14-20-26(35)18-12-13-30(3,4)38-29(18)23(28(20)37)21(32)11-10-17-8-6-5-7-9-17/h5-13,33-37H,14H2,1-4H3/b11-10+
Formula Weight 516.1784
Chemical Name or Material Rottlerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.