Learn More
OHAUS™ Rotor Swing Out ID Sealable
Product Code. 16662754 Shop All Ohaus Products
Click to view available options
Capacity:
4 x 750 mL
Unit Size:
Each
Product Code. 16662754
Brand: OHAUS™ 30553085
Offering Flexibility to Maximize Sample Preparation
  • Choose from a range of rotors to be used with Frontier centrifuges to match capacities and flexibility required for your basic to high-performance sample preparation needs.
  • Designed with usability in mind, the Frontier rotors can be effortlessly installed or removed in a few quick and simple steps, and users have easy access to the samples.
  • The rotors are durably constructed of high-quality aluminum alloy or polypropylene to last through demanding workflows - from autoclave sterilization to sub-zero temperatures.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 x 750 mL
Max. Speed 4,500,000 rpm