missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotigotine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Rotigotine |
| CAS | 99755-59-6 |
| Analyte or Component Names | Rotigotine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H25 N O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | (6S)-5,6,7,8-Tetrahydro-6-[propyl[2-(2-thienyl)ethyl]amino]-1-naphthalenol, (-)-N 0437, (S)-(-)-5,6,7,8-Tetrahydro-6-[propyl[2-(2-thienyl)ethyl]amino]-1-naphthalenol, (S)-(-)-Rotigotine, (S)-6-[Propyl[2-(thiophen-2-yl)ethyl]amino]-5,6,7,8-tetrahydronaphthalen-1-ol, N 0923, Neupro, Rotigotine, SPM 962 |
| InChI Formula | InChI=1S/C19H25NOS/c1-2-11-20(12-10-17-6-4-13-22-17)16-8-9-18-15(14-16)5-3-7-19(18)21/h3-7,13,16,21H,2,8-12,14H2,1H3/t16-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCN(CCc1cccs1)[C@H]2CCc3c(O)cccc3C2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction