missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rotigotine hydrochloride

Product Code. 18084992
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18084992 10 mg 10mg
18075822 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084992 Supplier Tocris Bioscience™ Supplier No. 3896/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dopamine D2/D3 agonist

Dopamine D2 and D3 receptor agonist. Ki values are 13 and 0.71 nM for D2 and D3 respectively. Also has significant affinity for 5-HT1A and adrenergic α2B receptors. Exhibits antiparkinsonian acitivity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125572-93-2
Molecular Formula C19H26ClNOS

Specifications

Quantity 10 mg
Formula Weight 351.93
Purity >99%
Molecular Weight (g/mol) 351.933
Chemical Name or Material Rotigotine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.