missing translation for 'onlineSavingsMsg'
Learn More

Nedaplatin, MedChemExpress

Product Code. 16471967
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16471967

Brand: Medchem Express HY13700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nedaplatin (NSC 375101D) is a derivative of cisplatin and DNA damage agent.

Nedaplatin (NSC 375101D) is a derivative of cisplatin and DNA damage agent.

Specifications

Chemical Name or Material Nedaplatin
Physical Form Powder
CAS 95734-82-0
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C2H8N2O3Pt
For Use With (Application) Cancer-programmed cell death
Synonym NSC 375101D
Solubility Information H2O : 13.6 mg/mL (44.86 mM; Need ultrasonic and warming; DMSO can inactivate Nedaplatin's activity) ∣DMF : 1 mg/mL (3.30 mM; Need ultrasonic; DMSO can inactivate Nedaplatin's activity)
SMILES O=C1C[O-][Pt+2](N)(N)[O-]1
Molecular Weight (g/mol) 303.18
Formula Weight 303.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.