Learn More
IKA™ ROTAVISC PT100.8 Temperature Sensor
Product Code. 17247539 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
IKA™ ROTAVISC Viscometer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17247539 IKA™ ROTAVISC Viscometer Each
1 options
Product Code. 17247539 Supplier IKA™ Supplier No. 0025000413
  • ONLINE EXCLUSIVE PRICE
    £64.15
    £61.00 / Pack of 1
    Save £3.15
    4% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The ROTAVISC PT100.8 Temperature Sensor for use with IKA™ ROTAVISC Rotational Viscometers.
  • Temperature sensor for IKA™ ROTAVISC Rotational Viscometers
  • Durable stainless steel design
  • 3 mm diameter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Temperature Probe (Immersion)
For Use With (Equipment) IKA™ ROTAVISC Viscometer
Material AISI 316Ti
Temperature Range (Metric) -20°C to +350°C