Learn More
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly
Product Code. 15540580 Shop All Buchi Products
Change view
Click to view available options
Material (Coating):
No Safety Coating
P+G Safety Coating
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Coating) Unit Size
15540580 P+G Safety Coating Each
15530580 No Safety Coating Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15540580 Supplier BUCHI Supplier No. 11062433
  • ONLINE EXCLUSIVE PRICE
    £1450.00
    £1376.00 / Pack of 1
    Save £74.00
    5% Off
View all promotions
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Vertical Condenser Glass Assembly for Rotavapor R-300, 1500 cm2. Assembly features a large top inlet for easy cleaning, connection for vapor temperature sensor, and inner glass tubing/plate. Includes a 1 liter receiving flask, tubing, and a ball joint clamp.
  • Includes receiving flask (1 L), tubing, and a ball joint clamp
  • Does NOT include evaporating flask, vacuum seal, vapor duct, and condenser holder
  • For use with Rotavapor R-300, R-2XX
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condensor
For Use With (Application) Rotary Evaporator
For Use With (Equipment) Rotavap™ R-300, R-210, and R-215
Includes Receiving Flask, Tubing, and a Ball Joint Clamp
Material (Coating) P+G Safety Coating
Volume (Metric) 1 L