Learn More
BUCHI Rotavapor R-300 with Diagonal Glass Assembly, Electronic Lift
The Rotavapor R-300 meets the highest expectations of convenience and versatility in rotary evaporation. A modular design allows the R-300 to be extended to a fully integrated system, with a central interface regulating each component.
£4075.00 - £7755.00
Specifications
Weight (Metric) | 13.5 kg |
---|---|
Voltage | 100/240 V |
Speed | 10 to 280 rpm |
For Use With (Application) | Rotary Evaporation |
Cooling Surface | 1500 cm2 |
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
15501090
|
BUCHI
11R300211A001 |
Uncoated | B-301 Heating Bath |
£4075.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15511090
|
BUCHI
11R300211A011 |
Uncoated | B-301 Heating Bath, Woulff Bottle |
£4250.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15541090
|
BUCHI
11R300211A041 |
Uncoated | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6335.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15521090
|
BUCHI
11R300211A021 |
Uncoated | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£5945.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15551090
|
BUCHI
11R300211A051 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£7425.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15531090
|
BUCHI
11R300211A031 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7035.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15551190
|
BUCHI
11R300251A011 |
Uncoated | B-305 Heating Bath, Woulff Bottle |
£4455.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15581190
|
BUCHI
11R300251A041 |
Uncoated | B-305 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6540.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15561190
|
BUCHI
11R300251A021 |
Uncoated | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6150.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15591190
|
BUCHI
11R300251A051 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£7630.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15571190
|
BUCHI
11R300251A031 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7240.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15561090
|
BUCHI
11R300211A101 |
P+G Safety Coating | B-301 Heating Bath |
£4195.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
15571090
|
BUCHI
11R300211A111 |
P+G Safety Coating | B-301 Heating Bath, Woulff Bottle |
£4380.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15501100
|
BUCHI
11R300211A141 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6460.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15581090
|
BUCHI
11R300211A121 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6080.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15511100
|
BUCHI
11R300211A151 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£7550.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15591090
|
BUCHI
11R300211A131 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7170.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15413224
|
BUCHI
11R300251A101 |
P+G Safety Coating | B-305 Heating Bath |
£4400.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15501200
|
BUCHI
11R300251A111 |
P+G Safety Coating | B-305 Heating Bath, Woulff Bottle |
£4585.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15531200
|
BUCHI
11R300251A141 |
P+G Safety Coating | B-305 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6675.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15511200
|
BUCHI
11R300251A121 |
P+G Safety Coating | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6285.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15541200
|
BUCHI
11R300251A151 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£7755.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
15521200
|
BUCHI
11R300251A131 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7375.00
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
Description
- A combination of convenience and versatility in rotary evaporation
- Modular design allows the R-300 to be extended to a fully integrated system
- Heating Bath B-305 can be used with either water or oil heating fluid
- Central interface compatibility to individually regulate each component
- User-friendly operation due to ergonomic electronic or manual lift options
Diagonal condenser
- For standard applications where height is limited
Smart Distillations
- Enables automated, unattended processes with optimized system parameters to take place
- Users can program their own SOPs into the system, then remotely monitor distillation processes
- BUCHI Cloud Services substantially maximize system uptimes with smart self-service tools and free updates
High Flexibility
- Modular design helps to meet individual requirements for rotary evaporation though high-level system flexibility
- A wide range of system accessories, glassware, and mobile monitoring options are available
- Users are capable of customizing their specific distillation process to fit any desired application
- Seven different glass assemblies that are compatible with various solvents and sample properties
Maximum Convenience
- The R-300 can be easily operated with features such as one-touch interface commands, and fast flask handling with Combi-Clip
- Combi-Clip with snap lock mechanism allows one-handed connection of evaporating flasks
- Options to manually adjust system parameters with intuitively placed buttons
- Immediate adjustment of heating temperature and rotation speed in the case of an urgent or unexpected event
- Exclusive possibility to personalize glassware with a customized permanent print
Specifications
13.5 kg | |
10 to 280 rpm | |
1500 cm2 | |
Diagonal | |
10° to 50° | |
Rotary Evaporator | |
Rotavapor R-300 |
100/240 V | |
Rotary Evaporation | |
100 W | |
IP 21 | |
CE/CSA Approved | |
50/60 Hz | |
29/32 |