Learn More
BUCHI Rotavapor R-300 with Cold Trap, Electronic Lift
The Rotavapor R-300 meets the highest expectations of convenience and versatility in rotary evaporation. A modular design allows the R-300 to be extended to a fully integrated system, with a central interface regulating each component.
£4485.00 - £8410.00
Specifications
Weight (Metric) | 13.5 kg |
---|---|
Voltage | 100/240 V |
Speed | 10 to 280 rpm |
For Use With (Application) | Rotary Evaporation |
Cooling Surface | 500 cm2 |
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
15521100
|
BUCHI
11R300211C001 |
Uncoated | B-301 Heating Bath |
£4485.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15531100
|
BUCHI
11R300211C011 |
Uncoated | B-301 Heating Bath, Woulff Bottle |
£4660.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15561100
|
BUCHI
11R300211C041 |
Uncoated | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6745.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15541100
|
BUCHI
11R300211C021 |
Uncoated | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6365.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15571100
|
BUCHI
11R300211C051 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£7835.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15551100
|
BUCHI
11R300211C031 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7455.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15551200
|
BUCHI
11R300251C001 |
Uncoated | B-305 Heating Bath |
£4680.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15561200
|
BUCHI
11R300251C011 |
Uncoated | B-305 Heating Bath, Woulff Bottle |
£4865.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15591200
|
BUCHI
11R300251C041 |
Uncoated | B-305 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6960.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15571200
|
BUCHI
11R300251C021 |
Uncoated | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6570.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15501210
|
BUCHI
11R300251C051 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8040.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15581200
|
BUCHI
11R300251C031 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7660.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15455865
|
BUCHI
11R300211C101 |
P+G Safety Coating | B-301 Heating Bath |
£4610.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15581100
|
BUCHI
11R300211C111 |
P+G Safety Coating | B-301 Heating Bath, Woulff Bottle |
£4795.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15511110
|
BUCHI
11R300211C141 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£6880.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15591100
|
BUCHI
11R300211C121 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6500.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15501110
|
BUCHI
11R300211C131 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7580.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15475865
|
BUCHI
11R300251C101 |
P+G Safety Coating | B-305 Heating Bath |
£4815.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15511210
|
BUCHI
11R300251C111 |
P+G Safety Coating | B-305 Heating Bath, Woulff Bottle |
£5000.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15541210
|
BUCHI
11R300251C141 |
P+G Safety Coating | B-305 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£7085.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15551210
|
BUCHI
11R300251C151 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8410.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15531210
|
BUCHI
11R300251C131 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£8015.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
15521110
|
BUCHI
11R300211C151 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15521210
|
BUCHI
11R300251C121 |
P+G Safety Coating | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- A combination of convenience and versatility in rotary evaporation
- Modular design allows the R-300 to be extended to a fully integrated system
- Heating Bath B-305 can be used with either water or oil heating fluid
- Central interface compatibility to individually regulate each component
- User-friendly operation due to ergonomic electronic or manual lift options
Cold Trap (Dry Ice) condenser
- For distillation of solvents with low boiling points
- Maximum condensation of vapors
- No cooling water is needed, but i.e. dry ice or ice
- Compatible with foam sensor
Smart Distillations
- Enables automated, unattended processes with optimized system parameters to take place
- Users can program their own SOPs into the system, then remotely monitor distillation processes
- BUCHI Cloud Services substantially maximize system uptimes with smart self-service tools and free updates
High Flexibility
- Modular design helps to meet individual requirements for rotary evaporation though high-level system flexibility
- A wide range of system accessories, glassware, and mobile monitoring options are available
- Users are capable of customizing their specific distillation process to fit any desired application
- Seven different glass assemblies that are compatible with various solvents and sample properties
Maximum Convenience
- The R-300 can be easily operated with features such as one-touch interface commands, and fast flask handling with Combi-Clip
- Combi-Clip with snap lock mechanism allows one-handed connection of evaporating flasks
- Options to manually adjust system parameters with intuitively placed buttons
- Immediate adjustment of heating temperature and rotation speed in the case of an urgent or unexpected event
- Exclusive possibility to personalize glassware with a customized permanent print
Specifications
13.5 kg | |
10 to 280 rpm | |
500 cm2 | |
Cold Trap | |
10° to 50° | |
Rotary Evaporator | |
Rotavapor™ R-300 |
100/240 V | |
Rotary Evaporation | |
100 W | |
IP 21 | |
CE/CSA Approved | |
50/60 Hz | |
29/32 |