missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Rosuvastatin calcium

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Rosuvastatin calcium is a potent HMG-CoA reductase inhibitor (IC50 = 5.4 nM). Inhibits cholesterol synthesis in rat hepatocytes in vitro and in vivo. Reduces plasma LDL cholesterol levels. Enhances lentiviral transduction of natural killer cells. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (3 R,5 S,6 E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl])-3,5-dihydroxyhept-6-enoic acid hemicalcium salt |
| CAS | 147098-20-2 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Target | HMG-CoA Reductase Inhibitors |
| Molecular Formula | C22H27FN3O6S.1⁄2Ca |
| Formula Weight | Observed MW: 500.57 |
| Purity | 99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction