missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin Calcium, MedChemExpress

Product Code. 16471266
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16471266

Brand: Medchem Express HY17504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rosuvastatin Calcium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM.

Rosuvastatin Calcium potently blocks hERG current with an IC50 of 195 nM. Rosuvastatin Calcium reduces the expression of the mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein. Rosuvastatin Calcium effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.

Specifications

Chemical Name or Material Rosuvastatin Calcium
CAS 147098-20-2
Color White
Purity Grade Notes Research
Molecular Formula C22H27Ca0.5FN3O6S
Quantity 200 mg
Synonym Rosuvastatin hemicalcium ZD 4522 Calcium
Solubility Information DMSO : 25 mg/mL (49.94 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.00 mM; Need ultrasonic)
SMILES O=C([O-])C[C@H](O)C[C@H](O)/C=C/C1=C(C(C)C)N=C(N(C)S(=O)(C)=O)N=C1C2=CC=C(F)C=C2.[0.5Ca2+]
Molecular Weight (g/mol) 500.57
Formula Weight 500.57
Percent Purity 96.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.