missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rosmarinic Acid, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Rosmarinic Acid
Specifications
Specifications
| Chemical Name or Material | Rosmarinic Acid |
| CAS | 20283-92-5 |
| Analyte or Component Names | Rosmarinic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H16 O8 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Rosmarinic Acid, Cinnamic acid, 3,4-dihydroxy-, 2-ester with 3-(3,4-dihydroxyphenyl)lactic acid, Rosmarinic acid, Labiatenic acid, Labiatic acid, Mamorekku RUH 2, Meiji Red Perilla Polyphenol, NPLC 0542, RM 21A, Rosemaric acid, Rosemary acid, trans-Rosmarinic acid, Benzenepropanoic acid, alpha-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, (alphaR)- (9CI), Benzenepropanoic acid, alpha-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, [R-(E)]- |
| InChI Formula | InChI=1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1 |
| SMILES | C([C@@H](OC(/C=C/C1=CC(O)=C(O)C=C1)=O)C(O)=O)C2=CC(O)=C(O)C=C2 |
| IUPAC Name | (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction