missing translation for 'onlineSavingsMsg'
Learn More

Rosmarinic acid, MedChemExpress

Product Code. 16409106
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409106 100 mg 100mg
16404857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16409106 Supplier Medchem Express Supplier No. HYN0529
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rosmarinic acid is a widespread phenolic ester compound in the plants. Rosmarinic acid inhibits MAO-A, MAO-B and COMT enzymes with IC50s of 50.1, 184.6 and 26.7 μM, respectively.

Rosmarinic acid is a widespread phenolic ester compound in the plants. Rosmarinic acid inhibits MAO-A, MAO-B and COMT enzymes with IC50s of 50.1, 184.6 and 26.7 μM, respectively.

Specifications

Chemical Name or Material Rosmarinic acid
CAS 20283-92-5
Color Light Brown
Purity Grade Notes Research
Molecular Formula C18H16O8
Quantity 100 mg
Synonym Labiatenic acid
Solubility Information DMSO : 62.5 mg/mL (173.46 mM; Need ultrasonic)
SMILES OC([C@H](OC(/C=C/C1=CC(O)=C(O)C=C1)=O)CC2=CC(O)=C(O)C=C2)=O
Molecular Weight (g/mol) 360.31
Formula Weight 360.31
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.