missing translation for 'onlineSavingsMsg'
Learn More

Rosmarinic Acid, 98.6%, MP Biomedicals™

Product Code. 11435881 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg

Product Code. 11435881

Brand: MP Biomedicals 0215979205

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 20283-92-5
Molecular Formula C18H16O8
Molecular Weight (g/mol) 360.318
MDL Number MFCD00017740
InChI Key DOUMFZQKYFQNTF-WUTVXBCWSA-N
Synonym rosmarinic acid, rosemary acid, r-rosmarinic acid, unii-mqe6xg29yi, rosmarinate, mqe6xg29yi, labiatenic acid, r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid, 3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester, 2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid
PubChem CID 5281792
ChEBI CHEBI:50371
IUPAC Name (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
SMILES C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O

Specifications

Color Beige
Assay Percent Range 0.986
Quantity 5 mg
Formula Weight 360.32
Physical Form Powder
Chemical Name or Material Rosmarinic acid, Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.