missing translation for 'onlineSavingsMsg'
Learn More

Rosmanol, MedChemExpress

Product Code. 16471748
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471748 5 mg 5mg
16459308 10 mg 10mg
16403966 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471748 Supplier Medchem Express Supplier No. HYN5015
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rosmanol could inhibit the oxidation of low density lipoprotein (LPL) and significantly inhibit lipopolysaccharide induced iNOS and COX-2 expression, with anti-inflammatory effect.

Rosmanol could inhibit the oxidation of low density lipoprotein (LPL) and significantly inhibit lipopolysaccharide induced iNOS and COX-2 expression, with anti-inflammatory effect.

Specifications

Chemical Name or Material Rosmanol
CAS 80225-53-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H26O5
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (288.67 mM; Need ultrasonic)
SMILES O=C1O[C@]2([H])[C@@H](O)C3=C(C(O)=C(O)C(C(C)C)=C3)[C@@]41CCCC(C)(C)[C@]24[H]
Molecular Weight (g/mol) 346.42
Formula Weight 346.42
Percent Purity 96.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.