missing translation for 'onlineSavingsMsg'
Learn More

Rosiptor, MedChemExpress

Product Code. 16440587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440587 50 mg 50mg
16424258 10 mg 10mg
16436977 5 mg 5mg
16445186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16440587 Supplier Medchem Express Supplier No. HY109011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rosiptor (AQX-1125) is a selective and orally active phosphatase SHIP1 activator with anti-inflammatory effects. Rosiptor (AQX-1125) inhibits Akt phosphorylation, inflammatory mediator production and leukocyte chemotaxis in vitro.

Rosiptor (AQX-1125) is a selective and orally active phosphatase SHIP1 activator with anti-inflammatory effects. Rosiptor (AQX-1125) inhibits Akt phosphorylation, inflammatory mediator production and leukocyte chemotaxis in vitro.

Specifications

Chemical Name or Material Rosiptor
CAS 782487-28-9
Color White
Purity Grade Notes Research
Molecular Formula C20H35NO2
Quantity 50 mg
Synonym AQX-1125
Solubility Information DMSO : 150 mg/mL (466.56 mM; Need ultrasonic and warming)
SMILES C[C@@]([C@H]1CO)(CC[C@H](O)C1)[C@](CC[C@@]2(C)[C@@]3([H])CCC2=C)([H])[C@H]3CN
Molecular Weight (g/mol) 321.5
Formula Weight 321.5
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.