missing translation for 'onlineSavingsMsg'
Learn More

Rosin

Product Code. 11304267
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11304267

Brand: Thermo Scientific Alfa Aesar 036734.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8050-09-7 | C20H30O2 | 302.46 g/mol

Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8050-09-7
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00132205
InChI Key RSWGJHLUYNHPMX-UHFFFAOYNA-N
Synonym 4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid

Specifications

Quantity 1 kg
Merck Index 14,8266
Solubility Information Soluble in alcohol, benzene, ether; Insoluble in water
Physical Form Powder or Solid
Chemical Name or Material Rosin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.