missing translation for 'onlineSavingsMsg'
Learn More

Rosiglitazone, MedChemExpress

Product Code. p-200096279
Encompass_Preferred
Click to view available options
Quantity:
50 mg
200 mg
Unit Size:
200mg
50mg
This item is not returnable. View return policy

Product Code. 16413337

Brand: Medchem Express HY17386

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rosiglitazone (BRL 49653) is an orally active selective PPARγ agonist (EC50: 60 nM, Kd: 40 nM). Rosiglitazone is an TRPC5 activator (EC50: 30 μM) and TRPM3 inhibitor. Rosiglitazone can be used in the research of obesity and diabetes, senescence, ovarian cancer.

Rosiglitazone (BRL 49653) is an orally active selective PPARγ agonist (EC50: 60 nM, Kd: 40 nM). Rosiglitazone is an TRPC5 activator (EC50: 30 μM) and TRPM3 inhibitor. Rosiglitazone can be used in the research of obesity and diabetes, senescence, ovarian cancer.

Specifications

Chemical Name or Material Rosiglitazone
CAS 122320-73-4
Color White
Purity Grade Notes Research
Molecular Formula C18H19N3O3S
Quantity 50 mg
Synonym BRL 49653
Solubility Information DMSO : 250 mg/mL (699.44 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (5.60 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(N1)SC(CC2=CC=C(OCCN(C)C3=NC=CC=C3)C=C2)C1=O
Molecular Weight (g/mol) 357.43
Formula Weight 357.43
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.