missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Rosiglitazone
Description
- Rosiglitazone is a potent and selective PPARγ ligand
- It binds to the PPARγ ligand-binding domain with a Kd of 43 nM (Lehmann et al.) It activates luciferase-based expression constructs PPARγ1 and PPARγ2 with EC50 values of approximately 30 nM and 100 nM, respectively (Lehmann et al.)
- Stimulates embryonic mouse neural stem cell proliferation and inhibits neuronal differentiation (Wada et al.)
- Induces adipocyte differentiation in C3H10T1/2 stem cells (Lehmann et al.)
- Promotes endothelial differentiation, and inhibits smooth muscle differentiation, in angiogenic progenitor cells (Wang et al.)
- Stimulates adipocyte differentiation, and decreases osteogenesis, in mouse and human bone marrow-derived mesenchymal stem cells (Ali et al.; Benvenuti et al.; Lecka-Czernik et al.; Sorocéanu et al.)
- Enhances osteoclast formation in mouse bone marrow cells stimulated with RANKL and M-CSF (Wu et al.)
Specifications
Specifications
| CAS | 122320-73-4 |
| Synonym | Avandia; BRL 49653 |
| Quantity | 50 mg |
| Molecular Formula | C18H19N3O3S |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?